COMH93D5FD23 Combi-Blocks, Inc.

(E)-Methyl 3-(3-oxocyclohex-1-en-1-yl)acrylate

  • MDL number MFCD30471959
  • Cas Number : 1280213-99-1
  • Molecular Weight : 180.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3893

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3893
Availability details for
Quantity
  • Availability
  • Add to Cart