COMH93D5FD38 Combi-Blocks, Inc.

2-Bromo-4-fluoro-1-(2-iodoethyl)benzene

  • MDL number MFCD30948571
  • Cas Number : 1804013-38-4
  • Molecular Weight : 328.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3920
Availability details for
Quantity
  • Availability
  • Add to Cart