COMH93D5FD53 Combi-Blocks, Inc.

4-Bromo-7-nitroisoquinoline

  • MDL number MFCD22200025
  • Cas Number : 347146-29-6
  • Molecular Weight : 253.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3955

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3955
Availability details for
Quantity
  • Availability
  • Add to Cart