COMH93D5FD69 Combi-Blocks, Inc.

6-Oxabicyclo[3.1.0]hexan-2-one

  • MDL number MFCD24687916
  • Cas Number : 6705-52-8
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3988

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-3988
Availability details for
Quantity
  • Availability
  • Add to Cart