COMH93D5FD7E Combi-Blocks, Inc.

4-[Bis(9,9-dimethylfluoren-2-yl)amino]azobenzene

  • MDL number MFCD29089342
  • Cas Number : 883554-70-9
  • Molecular Weight : 581.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4015

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4015
Availability details for
Quantity
  • Availability
  • Add to Cart