COMH93D5FD8C Combi-Blocks, Inc.

(N-(Phenylmethoxy)carbonyl)-n-(phenylmethyl)-glycine

  • MDL number MFCD30595703
  • Cas Number : 71922-59-3
  • Molecular Weight : 299.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4033

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4033
Availability details for
Quantity
  • Availability
  • Add to Cart