COMH93D5FD96 Combi-Blocks, Inc.

Racemic-(1s,4s)-tert-butyl 1-vinyl-2-oxa-5-azabicyclo[2.2.1]heptane-5-carboxylate

  • MDL number MFCD31579966
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4045
Availability details for
Quantity
  • Availability
  • Add to Cart