COMH93D5FDA3 Combi-Blocks, Inc.

(3-Cyano-4-isobutoxyphenyl)boronic acid

  • MDL number MFCD26387475
  • Cas Number : 876918-32-0
  • Molecular Weight : 219.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4063

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4063
Availability details for
Quantity
  • Availability
  • Add to Cart