COMH93D5FDA4 Combi-Blocks, Inc.

2-(3-(Trifluoromethoxy)phenoxy)ethanol

  • MDL number MFCD27949210
  • Cas Number : 313655-84-4
  • Molecular Weight : 222.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4065

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4065
Availability details for
Quantity
  • Availability
  • Add to Cart