COMH93D5FDA6 Combi-Blocks, Inc.

1-(4-Butyl-phenyl)-3-piperidin-3-yl-urea

  • MDL number MFCD30855935
  • Cas Number : 2197648-37-4
  • Molecular Weight : 275.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4067
Availability details for
Quantity
  • Availability
  • Add to Cart