COMH93D5FDB1 Combi-Blocks, Inc.

Methyl 6-methyl-1h-indazole-5-carboxylate

  • MDL number MFCD23705407
  • Cas Number : 1427405-21-7
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4082

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4082
Availability details for
Quantity
  • Availability
  • Add to Cart