COMH93D5FDD3 Combi-Blocks, Inc.

1-Ethylcyclopentyl methacrylate

  • MDL number MFCD23703079
  • Cas Number : 266308-58-1
  • Molecular Weight : 182.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4123

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4123
Availability details for
Quantity
  • Availability
  • Add to Cart