COMH93D5FDFE Combi-Blocks, Inc.

4-(2-Fluoropyridin-3-yl)-2-(methylthio)pyrimidine

  • MDL number MFCD31652323
  • Cas Number : 945375-80-4
  • Molecular Weight : 221.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4177

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4177
Availability details for
Quantity
  • Availability
  • Add to Cart