COMH93D5FE24 Combi-Blocks, Inc.

5,5,10,10,15,15-Hexamethyl-10,15-dihydro-5h-diindeno[1,2-a:1',2'-c]fluorene

  • MDL number MFCD26793578
  • Cas Number : 597554-76-2
  • Molecular Weight : 426.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4223

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4223
Availability details for
Quantity
  • Availability
  • Add to Cart