COMH93D5FE25 Combi-Blocks, Inc.

(R)-3,3-Difluoro-8-azaspiro[4.5]decan-1-amine 2hcl

  • MDL number MFCD30530882
  • Cas Number : 2055849-17-5
  • Molecular Weight : 263.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4224
Availability details for
Quantity
  • Availability
  • Add to Cart