COMH93D5FF54 Combi-Blocks, Inc.

2-Methoxy-4,6-dimethylbenzaldehyde

  • MDL number MFCD22415104
  • Cas Number : 51926-66-0
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4596

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4596
Availability details for
Quantity
  • Availability
  • Add to Cart