COMH93D5FFAD Combi-Blocks, Inc.

4-(3-(Bromomethyl)phenyl)morpholine hbr

  • MDL number MFCD29921206
  • Cas Number : 1956370-51-6
  • Molecular Weight : 337.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4713

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4713
Availability details for
Quantity
  • Availability
  • Add to Cart