COMH93D6003A Combi-Blocks, Inc.

2-Chloro-4-thiophen-3-yl-thieno[3,2-d]pyrimidine

  • MDL number MFCD28675738
  • Cas Number : 443149-08-4
  • Molecular Weight : 252.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4888

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4888
Availability details for
Quantity
  • Availability
  • Add to Cart