COMH93D60053 Combi-Blocks, Inc.

Fmoc-l-cys-otbu

  • MDL number MFCD28347836
  • Molecular Weight : 399.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-4919
Availability details for
Quantity
  • Availability
  • Add to Cart