COMH93D600AC Combi-Blocks, Inc.

1-Phenyl-2,5,8,11,14,17,20,23,26,29-decaoxahentriacontan-31-ol

  • MDL number MFCD24682798
  • Cas Number : 908258-44-6
  • Molecular Weight : 548.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5030

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5030
Availability details for
Quantity
  • Availability
  • Add to Cart