COMH93D600C4 Combi-Blocks, Inc.

3-(Methoxycarbonyl)-5-propoxybenzoic acid

  • MDL number MFCD27930758
  • Cas Number : 869529-27-1
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5062

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5062
Availability details for
Quantity
  • Availability
  • Add to Cart