COMH93D60178 Combi-Blocks, Inc.

Acid yellow 220

  • MDL number MFCD22200817
  • Cas Number : 70851-34-2
  • Molecular Weight : 1206.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5278

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5278
Availability details for
Quantity
  • Availability
  • Add to Cart