COMH93D60191 Combi-Blocks, Inc.

N-(4-Chlorobenzyl)-n-(2,2-dimethoxyethyl)-4-methylbenzenesulfonamide

  • MDL number MFCD28402185
  • Cas Number : 54879-43-5
  • Molecular Weight : 383.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5312

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5312
Availability details for
Quantity
  • Availability
  • Add to Cart