COMH93D601F4 Combi-Blocks, Inc.

3-(Pyridin-4-yl)-4,5,6,7-tetrahydroisoxazolo[4,5-c]pyridine

  • MDL number MFCD24614574
  • Cas Number : 1190971-47-1
  • Molecular Weight : 201.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5434

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5434
Availability details for
Quantity
  • Availability
  • Add to Cart