COMH93D60203 Combi-Blocks, Inc.

(2-Benzyl-9-fluoro-1,2,3,3a,4,9b-hexahydrochromeno[3,4-c]pyrrol-3a-yl)methanol

  • MDL number MFCD31555264
  • Cas Number : 2102411-27-6
  • Molecular Weight : 313.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5451

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5451
Availability details for
Quantity
  • Availability
  • Add to Cart