COMH93D6023F Combi-Blocks, Inc.

N-[2-(3,4-Dimethoxyphenyl)acetyl]-n'-(tert-butyloxycarbonyl)-guanidine

  • MDL number MFCD31654495
  • Cas Number : 1620554-49-5
  • Molecular Weight : 337.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5525

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5525
Availability details for
Quantity
  • Availability
  • Add to Cart