COMH93D60266 Combi-Blocks, Inc.

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2,6-difluorophenyl)-2-methylpropanoic acid

  • MDL number MFCD22989428
  • Cas Number : 1223105-51-8
  • Molecular Weight : 437.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5570
Availability details for
Quantity
  • Availability
  • Add to Cart