COMH93D602AF Combi-Blocks, Inc.

(R)-3-Methylpiperidine-3-carboxylic acid hcl

  • MDL number MFCD30828734
  • Molecular Weight : 179.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5659

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5659
Availability details for
Quantity
  • Availability
  • Add to Cart