COMH93D602D5 Combi-Blocks, Inc.

5-Bromo-1,2,3-trimethylbenzene

  • MDL number MFCD28746982
  • Cas Number : 32591-43-8
  • Molecular Weight : 199.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5704

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5704
Availability details for
Quantity
  • Availability
  • Add to Cart