COMH93D602F4 Combi-Blocks, Inc.

1-(3-Aminoquinolin-5-yl)ethanone

  • MDL number MFCD30828912
  • Cas Number : 2089649-29-4
  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5746

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5746
Availability details for
Quantity
  • Availability
  • Add to Cart