COMH93D60311 Combi-Blocks, Inc.

1-Benzhydryl-n,n,3-trimethylazetidin-3-amine

  • MDL number MFCD27998387
  • Cas Number : 133891-61-9
  • Molecular Weight : 280.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5784

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5784
Availability details for
Quantity
  • Availability
  • Add to Cart