COMH93D60368 Combi-Blocks, Inc.

Methyl 2-(1-benzyl-1h-pyrazol-4-yl)acetate

  • MDL number MFCD28135420
  • Cas Number : 1823527-33-8
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5889

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5889
Availability details for
Quantity
  • Availability
  • Add to Cart