COMH93D60389 Combi-Blocks, Inc.

2-Chloro-4-morpholin-4-yl-7-(2,2,2-trifluoroethyl)-7h-pyrrolo[ 2,3-d]pyrimidine

  • MDL number MFCD28383661
  • Cas Number : 1202885-79-7
  • Molecular Weight : 320.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-5930
Availability details for
Quantity
  • Availability
  • Add to Cart