COMH93D603C6 Combi-Blocks, Inc.

(1R,4S,5R)-Rel-5-acetyl-1-boc-2-azabicyclo[2.1.1]hexane

  • MDL number MFCD28501243
  • Cas Number : 615575-71-8
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6008

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6008
Availability details for
Quantity
  • Availability
  • Add to Cart