COMH93D603C8 Combi-Blocks, Inc.

(6-(Trifluoromethyl)pyridin-3-yl)boronic acid hydrochloride

  • MDL number MFCD27965243
  • Cas Number : 2304634-22-6
  • Molecular Weight : 227.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6011

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6011
Availability details for
Quantity
  • Availability
  • Add to Cart