COMH93D603E3 Combi-Blocks, Inc.

1,2-Benzenediol,4-octyl-

  • MDL number MFCD30342423
  • Cas Number : 7580-46-3
  • Molecular Weight : 222.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6041

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6041
Availability details for
Quantity
  • Availability
  • Add to Cart