COMH93D60448 Combi-Blocks, Inc.

((2R,5S)-5-(4-Amino-5-fluoro-2-oxopyrimidin-1(2h)-yl)-1,3-oxathiolan-2-yl)methyl butyrate

  • MDL number MFCD30185762
  • Cas Number : 143491-56-9
  • Molecular Weight : 317.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6168

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6168
Availability details for
Quantity
  • Availability
  • Add to Cart