COMH93D6044F Combi-Blocks, Inc.

(5-tert-Butyl-1,3-phenylene)dimethanol

  • MDL number MFCD29489970
  • Cas Number : 22157-91-1
  • Molecular Weight : 194.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6179

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6179
Availability details for
Quantity
  • Availability
  • Add to Cart