COMH93D60472 Combi-Blocks, Inc.

(R)-5-(1-Phenylethyl)-5-azaspiro[2.4]heptan-7-one

  • MDL number MFCD29059427
  • Cas Number : 1956435-39-4
  • Molecular Weight : 215.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6224

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6224
Availability details for
Quantity
  • Availability
  • Add to Cart