COMH93D6048F Combi-Blocks, Inc.

2-Bromo-2-methylpropan-1-amine hbr

  • MDL number MFCD30828732
  • Cas Number : 36565-68-1
  • Molecular Weight : 232.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6261

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6261
Availability details for
Quantity
  • Availability
  • Add to Cart