COMH93D6049B Combi-Blocks, Inc.

3-Amino-n-(2-hydroxyethyl)-5-methylbenzamide

  • MDL number MFCD26855830
  • Cas Number : 1566647-80-0
  • Molecular Weight : 194.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6275

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6275
Availability details for
Quantity
  • Availability
  • Add to Cart