COMH93D604B7 Combi-Blocks, Inc.

Tetrahydrothiopyran-4-ylboronic acid

  • MDL number MFCD31619561
  • Cas Number : 2265921-88-6
  • Molecular Weight : 146.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6310

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6310
Availability details for
Quantity
  • Availability
  • Add to Cart