COMH93D604D5 Combi-Blocks, Inc.

2-(3-Isopropoxy-benzyl)-butyric acid

  • MDL number MFCD31583264
  • Cas Number : 1779839-89-2
  • Molecular Weight : 236.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6345

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6345
Availability details for
Quantity
  • Availability
  • Add to Cart