COMH93D604F8 Combi-Blocks, Inc.

4-(2-Bromoethynyl)benzoic acid

  • MDL number MFCD28252185
  • Cas Number : 1485684-77-2
  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6391

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6391
Availability details for
Quantity
  • Availability
  • Add to Cart