COMH93D604FF Combi-Blocks, Inc.

2-Chloro-6-methyl-4-(piperidine-1-carbonyl)pyridine

  • MDL number MFCD26431110
  • Molecular Weight : 238.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6398

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6398
Availability details for
Quantity
  • Availability
  • Add to Cart