COMH93D60504 Combi-Blocks, Inc.

2-Acetyl-6-bromo-8-cyclopentyl-5-methylpyrido[2,3-d]pyrimidin-7(8h)-one

  • MDL number MFCD30730141
  • Cas Number : 2102411-08-3
  • Molecular Weight : 350.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6405

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6405
Availability details for
Quantity
  • Availability
  • Add to Cart