COMH93D60512 Combi-Blocks, Inc.

4-Nitro-2,3-dihydro-1h-inden-1-amine hydrochloride

  • MDL number MFCD30541452
  • Cas Number : 1958100-54-3
  • Molecular Weight : 214.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6422

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6422
Availability details for
Quantity
  • Availability
  • Add to Cart