COMH93D60584 Combi-Blocks, Inc.

Ethyl 3-bromo-7-methylpyrazolo[1,5-a]pyrimidine-5-carboxylate

  • MDL number MFCD27992241
  • Cas Number : 1823500-24-8
  • Molecular Weight : 284.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6574

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6574
Availability details for
Quantity
  • Availability
  • Add to Cart