COMH93D60597 Combi-Blocks, Inc.

5-Bromo-2-fluoro-3-iodobenzonitrile

  • MDL number MFCD30829168
  • Cas Number : 1131632-57-9
  • Molecular Weight : 325.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6601

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6601
Availability details for
Quantity
  • Availability
  • Add to Cart