COMH93D605B2 Combi-Blocks, Inc.

[4-(Methoxy-methyl-carbamoyl)-cyclohexylmethyl]-carbamic acid tert-butyl ester

  • MDL number MFCD24447778
  • Molecular Weight : 300.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6633

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?HD-6633
Availability details for
Quantity
  • Availability
  • Add to Cart